EMAIL THIS PAGE TO A FRIEND
Sensors (Basel, Switzerland)
Flow injection chemiluminescent immunoassay for carcinoembryonic antigen using boronic immunoaffinity column.
PMID 22303179
Abstract
A flow injection chemiluminescence immunoassay for rapid and sensitive detection of carcinoembryonic antigen (CEA) by using a phenylboronic acid-based immunoaffinity column as a glycoprotein collector was proposed in this paper. The column was prepared by coupling of 3-aminophenylboronic acid on the glass beads through a γ-glycidoxypropyltrimethoxysilane (GPMS) linkage. Based on an indirect competitive immunoreaction, the mixture of CEA sample and enzyme conjugated CEA antibody (HRP-anti-CEA) was incubated in advance, followed by direct injection to the column to capture free HRP-labeled CEA antibody in the column. The trapped HRP-labeled antibody was detected by flow inject chemiluminescence in the presence of luminol and hydrogen peroxide. The decreased chemiluminescent signal was proportional to the concentration of CEA in the range of 3.0-30.0 ng/mL with a correlation coefficient of 0.998. The column showed an acceptable reproducibility and stability and is potentially used for practical clinical detection of the serum CEA level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS003827
3-Aminophenylboronic acid, AldrichCPR
C6H8BNO2