EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Heart and circulatory physiology
Cytochrome P-450 2C9 exerts a vasoconstrictor influence on coronary resistance vessels in swine at rest and during exercise.
PMID 22307673
Abstract
A significant endothelium-dependent vasodilation persists after inhibition of nitric oxide synthase (NOS) and cyclooxygenase (COX) in the coronary vasculature, which has been linked to the activation of cytochrome P-450 (CYP) epoxygenases expressed in endothelial cells and subsequent generation of vasodilator epoxyeicosatrienoic acids. Here, we investigated the contribution of CYP 2C9 metabolites to regulation of porcine coronary vasomotor tone in vivo and in vitro. Twenty-six swine were chronically instrumented. Inhibition of CYP 2C9 with sulfaphenazole (5 mg/kg iv) alone had no effect on bradykinin-induced endothelium-dependent coronary vasodilation in vivo but slightly attenuated bradykinin-induced vasodilation in the presence of combined NOS/COX blockade with N(ω)-nitro-L-arginine (20 mg/kg iv) and indomethacin (10 mg/kg iv). Sulfaphenazole had minimal effects on coronary resistance vessel tone at rest or during exercise. Surprisingly, in the presence of combined NOS/COX blockade, a significant coronary vasodilator response to sulfaphenzole became apparent, both at rest and during exercise. Subsequently, we investigated in isolated porcine coronary small arteries (∼250 μm) the possible involvement of reactive oxygen species (ROS) in the paradoxical vasoconstrictor influence of CYP 2C9 activity. The vasodilation by bradykinin in vitro in the presence of NOS/COX blockade was markedly potentiated by sulfaphenazole under control conditions but not in the presence of the ROS scavenger N-(2-mercaptoproprionyl)-glycine. In conclusion, CYP 2C9 can produce both vasoconstrictor and vasodilator metabolites. Production of these metabolites is enhanced by combined NOS/COX blockade and is critically dependent on the experimental conditions. Thus production of vasoconstrictors slightly outweighed the production of vasodilators at rest and during exercise. Pharmacological stimulation with bradykinin resulted in vasodilator CYP 2C9 metabolite production when administered in vivo, whereas vasoconstrictor CYP 2C9 metabolites, most likely ROS, were dominant when administered in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6635
N-(2-Mercaptopropionyl)glycine
C5H9NO3S