EMAIL THIS PAGE TO A FRIEND
Molecular endocrinology (Baltimore, Md.)
Endothelin-converting enzyme-1 actions determine differential trafficking and signaling of corticotropin-releasing factor receptor 1 at high agonist concentrations.
PMID 22322595
Abstract
CRF receptor 1 (CRF(1)), a key neuroendocrine mediator of the stress response, has two known agonists corticotropin-releasing factor (CRF) and urocortin 1 (Ucn1). Here we report that endothelin-converting enzyme-1 (ECE-1) differentially degrades CRF and Ucn1; ECE-1 cleaves Ucn1, but not CRF, at critical residue Arginine-34/35', which is essential for ligand-receptor binding. At near K(D) agonist concentration (30 nm), both Ucn1- and CRF-mediated Ca(2+) mobilization are ECE-1 dependent. Interestingly, at high agonist concentration (100 nm), Ucn1-mediated Ca(2+) mobilization remains ECE-1 dependent, whereas CRF-mediated mobilization becomes independent of ECE-1 activity. At high agonist concentration, ECE-1 inhibition disrupted Ucn1-, but not CRF-induced CRF(1) recycling and resensitization, but did not prolong the association of CRF(1) with β-arrestins. RNA interference-mediated knockdown of Rab suggests that both Ucn1- and CRF-induced CRF(1) resensitization is dependent on activity of Rab11, but not of Rab4. CRF(1) behaves like a class A G protein-coupled receptor with respect to transient β-arrestins interaction. We propose that differential degradation by ECE-1 is a novel mechanism by which CRF(1) receptor is protected from overactivation by physiologically relevant high concentrations of higher affinity ligand to mediate distinct resensitization and downstream signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SCP0131 ECE-1 Substrate, ≥95% (HPLC)
C66H81N15O19