EMAIL THIS PAGE TO A FRIEND
Anti-cancer agents in medicinal chemistry
Gambogic acid is a novel anti-cancer agent that inhibits cell proliferation, angiogenesis and metastasis.
PMID 22339063
Abstract
Gambogic acid (GA) is a caged xanthone that is derived from Garcinia hanburyi and functions as a strong apoptotic inducer in many types of cancer cells. The distinct effectiveness of GA has led to its characterization as a novel anti-cancer agent. There is an increasing number of research studies focused on elucidating the molecular mechanisms of GA-induced anti-cancer effects, and several critical signaling pathways have been reported to be influenced by GA treatment. In this review, we summarize the multiple functional effects of GA administration in cancer cells including the induction of apoptosis, the inhibition of proliferation and the prevention of cancer metastasis and tumor angiogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8171
Gambogic acid, ≥95% (HPLC), powder
C38H44O8