EMAIL THIS PAGE TO A FRIEND
The Medical letter on drugs and therapeutics
In brief: etonogestrel (nexplanon) contraceptive implant.
PMID 22354222
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0356
Etonogestrel, ≥98% (HPLC)
C22H28O2