EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Effect of polyanetholesulphonic acid and xylan sulphate on antithrombin III activity.
PMID 223567
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2008
Polyanetholesulfonic acid sodium salt, powder