EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Sesquiterpenoids from Atractylodes macrocephala act as farnesoid X receptor and progesterone receptor modulators.
PMID 22365756
Abstract
Two sesquiterpenoids, atractylenolide II and III, were isolated and identified from Atractylodes macrocephala (Asteraceae) to be subsequently evaluated for their activity against farnesoid X receptor (FXR) and progesterone receptor (PR) by transient transfection reporter assays. These sesquiterpenoids did not exert significant agonistic effect but antagonized the activity of chenodeoxycholic acid (CDCA), an endogenous FXR agonist, for FXR driven SHP promoter transactivation. Additionally, they transactivated CYP7A1 gene promoter activity by antagonizing FXR. Apart from acting as a FXR antagonist, atractylenolide III also showed agonistic activity against PR. All these results demonstrated that atractylenolide II and III are the active components of Atractylodes macrocephala to exert specific pharmacologic effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2987
Atractylenolide III, ≥98% (HPLC)
C15H20O3