EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Subcellular localization and putative role of VPS13A/chorein in dopaminergic neuronal cells.
PMID 22366033
Abstract
Chorea-acanthocytosis (ChAc) is a rare hereditary neurodegenerative disorder caused by loss of function mutations in the vacuolar protein sorting 13 homolog A (VPS13A) gene encoding chorein. Although a deficiency in chorein function leads to apoptosis of striatal neurons in ChAc model mouse, its detailed subcellular localization and physiological role remain unclear. In this study, we produced two anti-chorein polyclonal antibodies and examined the intracellular localization of endogenous chorein in neuronal cells. Immunocytochemically, chorein was observed in the termini of extended neurites and partially colocalized with synaptotagmin I in differentiated PC12 cells. Subcellular localization analysis by sucrose density gradient fractionation showed that chorein and synaptotagmin I were located in dense-core vesicles (DCVs), which contain dopamine. In addition, PC12 cells stably expressing carboxyterminal fragment of chorein increased K(+)-induced dopamine release. Taken together, these results suggest that chorein is involved in exocytosis of DCV.