EMAIL THIS PAGE TO A FRIEND
Acta pharmacologica Sinica
Chronic cardiotoxicity of doxorubicin involves activation of myocardial and circulating matrix metalloproteinases in rats.
PMID 22447222
Abstract
To investigate the role of matrix metalloproteinases (MMPs) in the responses of rats to a prolonged doxorubicin (DOX) treatment. Male Wistar rats were used. DOX was administered by intraperitoneal injections of seven doses (cumulative dose was 15 mg/kg). Control animals were treated with saline. Tissue or plasma samples were collected at four and eight weeks after the application of the last dose. Protein levels were determined by immunoblot assay, and MMP activities were measured by gelatin zymography. Superoxide content was analyzed using a lucigenin chemiluminescence assay and superoxide dismutase (SOD) activities with a SOD assay kit. Qualitative structural alterations of the heart were characterized by transmission electron microscopy. Systolic blood pressure was higher in DOX-treated rats as compared with the control rats at 8 weeks after treatment. In contrast, there were no differences in the heart rate between the control and DOX-treated rats. DOX treatment caused marked heterogeneous subcellular alterations of cardiomyocytes and structural disorganizations of the cardiac extracellular space. The effects of DOX were linked to a stimulation of plasma MMP-2 and MMP-9 activities that had already increased by 4 weeks after the end of the treatment. In the left ventricle, however, DOX only led to increased MMP-2 activation at 8 weeks after the end of treatment. These changes in tissue MMP-2 were connected with stimulation of Akt kinase activation, inhibition of SOD, an increase in superoxide levels, induction of iNOS protein expression and caspase-3 activation. Our results show that MMPs are involved in the chronic cardiotoxicity of DOX in rats. The data also suggest that reactive oxygen species (superoxide), NO production (iNOS) and the Akt kinase pathway can modulate MMP-2 activities in rat hearts influenced by DOX.