EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Pseudoephenamine: a practical chiral auxiliary for asymmetric synthesis.
PMID 22461381
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
287636
(1S,2S)-(+)-Pseudoephedrine, 98%
C10H15NO