EMAIL THIS PAGE TO A FRIEND
Profiles of drug substances, excipients, and related methodology
Amlodipine besylate.
PMID 22469316
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1029501
Amlodipine besylate, United States Pharmacopeia (USP) Reference Standard
C20H25ClN2O5 · C6H5SO3H
A5605
Amlodipine besylate, ≥98% (HPLC)
C20H25ClN2O5 · C6H5SO3H