EMAIL THIS PAGE TO A FRIEND
The Medical letter on drugs and therapeutics
Low-dose sublingual zolpidem (Intermezzo) for insomnia due to middle-of-the-night awakening.
PMID 22469649
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z103
Zolpidem, ≥98% (HPLC), solid
C19H21N3O