EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Ligand effects on the stereochemical outcome of Suzuki-Miyaura couplings.
PMID 22480237
Abstract
The ligands associated with various Pd catalysts play a crucial role in determining the stereochemistry of cross-couplings between boronic acids and Z-alkenyl halides. A ligand on palladium has been found that leads to the desired products under mild conditions and in high yields that, in most cases, retain their Z-olefin geometry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
436836
2-Thienylboronic acid, ≥95.0%
C4H5BO2S