EMAIL THIS PAGE TO A FRIEND
Journal of emergency nursing: JEN : official publication of the Emergency Department Nurses Association
A review of epinephrine administration in pediatric anaphylaxis.
PMID 22494791