EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Towards a green process for bulk-scale synthesis of ethyl acetate: efficient acceptorless dehydrogenation of ethanol.
PMID 22517628