EMAIL THIS PAGE TO A FRIEND
Bioorganic chemistry
Decarboxylation mechanisms in biological system.
PMID 22534166
Abstract
This review examines the mechanisms propelling cofactor-independent, organic cofactor-dependent and metal-dependent decarboxylase chemistry. Decarboxylation, the removal of carbon dioxide from organic acids, is a fundamentally important reaction in biology. Numerous decarboxylase enzymes serve as key components of aerobic and anaerobic carbohydrate metabolism and amino acid conversion. In the past decade, our knowledge of the mechanisms enabling these crucial decarboxylase reactions has continued to expand and inspire. This review focuses on the organic cofactors biotin, flavin, NAD, pyridoxal 5'-phosphate, pyruvoyl, and thiamin pyrophosphate as catalytic centers. Significant attention is also placed on the metal-dependent decarboxylase mechanisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8754
Thiamine pyrophosphate, ≥95%
C12H19ClN4O7P2S