EMAIL THIS PAGE TO A FRIEND
Tetrahedron letters
Diversity Oriented Synthesis: Concise Entry to Novel Derivatives of Yohimbine and Corynanthe Alkaloids.
PMID 22544982
Abstract
A novel MCAP-cycloaddition sequence has been applied to the facile synthesis of β-carboline intermediates to gain rapid access to novel derivatives of yohimbine-like and corynanthe-like compounds that may be easily diversified by cross-coupling reactions and N-derivatizations to generate small compound libraries.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
473723
7-Bromoindole, 96%
C8H6BrN