EMAIL THIS PAGE TO A FRIEND
Anais brasileiros de dermatologia
Contact sensitivity to metals (chromium, cobalt and nickel) in childhood.
PMID 22570032
Abstract
Metals, especially nickel, are the most common contact allergens in children. Recent data has shown increased incidence of allergy in industrialized countries. Sensitization can occur at any age, even in neonates. Costume jewelry, particularly earrings, is linked to increased sensitization to nickel. Sensitization to cobalt often occurs by the use of costume jewelry. The most common source of sensitization to chromium is leather. Due to the absence of a specific therapy, the main treatment is to identify and avoid the responsible allergens. This article presents an updated view on the epidemiological and clinical aspects of contact allergy to metals, focusing on prevention strategies and risk factors, and warns about possible and new sources of contact.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF94594450
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
Co
GF41541476
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Co
GF45372315
Cobalt, foil, 4mm disks, thickness 0.003mm, 99.9%
Co
GF08560289
Cobalt, foil, 4mm disks, thickness 0.004mm, 99.9%
Co
GF89559343
Cobalt, foil, 4mm disks, thickness 0.005mm, 99.9%
Co
GF48392051
Cobalt, foil, 4mm disks, thickness 0.006mm, 99.9%
Co
GF55954873
Cobalt, foil, 4mm disks, thickness 0.007mm, 99.9%
Co
GF92926727
Cobalt, foil, 4mm disks, thickness 0.008mm, 99.9%
Co
GF70445246
Cobalt, foil, 4mm disks, thickness 0.009mm, 99.9%
Co
GF40089225
Cobalt, foil, 4mm disks, thickness 0.0125mm, 99.9%
Co
GF48813723
Cobalt, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Co
GF10782121
Cobalt, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
Co
GF40808516
Cobalt, foil, 100x100mm, thickness 0.25mm, as rolled, 99.9%
Co
GF18093461
Cobalt, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Co
GF30160291
Cobalt, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
Co
GF81966224
Cobalt, foil, 100x100mm, thickness 0.7mm, annealed, 99.9%
Co
GF56494187
Cobalt, foil, 100x100mm, thickness 1.0mm, as rolled, 99.9%
Co
GF76259593
Cobalt, foil, 100x100mm, thickness 2.0mm, as rolled, 99.9%
Co
GF91564904
Cobalt, foil, 250x250mm, thickness 0.50mm, annealed, 99.9%
Co
GF88288699
Cobalt, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Co
GF30959411
Cobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Co
GF67617153
Cobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Co
GF85563535
Cobalt, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Co
GF09035647
Cobalt, foil, 25x25mm, thickness 0.7mm, annealed, 99.9%
Co
GF41834011
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.9%
Co
GF34805268
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Co
GF40133472
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.9%
Co
GF03773640
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.9%
Co
GF06754406
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Co
GF42159315
Cobalt, foil, light tested, 25x25mm, thickness 0.009mm, 99.9%
Co