EMAIL THIS PAGE TO A FRIEND
Journal of biomedicine & biotechnology
Synthesis of 1,5-benzodiazepine and its derivatives by condensation reaction using H-MCM-22 as catalyst.
PMID 22570531
Abstract
A simple and versatile method for the synthesis of 1,5-benzodiazepines is via condensation of o-phenylenediamines (OPDA) and ketones in the presence of catalytic amount of H-MCM-22 using acetonitrile as solvent at room temperature. In all the cases, the reactions are highly selective and are completed within 1-3 h. The method is applicable to both cyclic and acyclic ketones without significant differences. The reaction proceeds efficiently under ambient conditions with good-to-excellent yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H10401
Hexamethyleneimine, 99%
C6H13N