EMAIL THIS PAGE TO A FRIEND
Journal of human genetics
Atypical hemolytic uremic syndrome and genetic aberrations in the complement factor H-related 5 gene.
PMID 22622361
Abstract
Atypical hemolytic uremic syndrome (aHUS) is a severe renal disorder that is associated with mutations in genes encoding proteins of the alternative complement pathway. Previously, we identified pathogenic variations in genes encoding complement regulators (CFH, CFI and MCP) in our aHUS cohort. In this study, we screened for mutations in the alternative pathway regulator CFHR5 in 65 aHUS patients by means of PCR on genomic DNA and sequence analysis. Potential pathogenicity of genetic alterations was determined by published data on CFHR5 variants, evolutionary conservation and in silico mutation prediction programs. Detection of serum CFHR5 was performed by western blot analysis and enzyme-linked immunosorbent assay. A potentially pathogenic sequence variation was found in CFHR5 in three patients (4.6%). All variations were located in short consensus repeats that might be involved in binding to C3b, heparin or C-reactive protein. The identified CFHR5 mutations require functional studies to determine their relevance to aHUS, but they might be candidates for an altered genetic profile predisposing to the disease.