EMAIL THIS PAGE TO A FRIEND
Journal of virology
Replication of minute virus of mice in murine cells is facilitated by virally induced depletion of p21.
PMID 22623787
Abstract
The DNA damage response to infection with minute virus of mice (MVM) leads to activated p53; however, p21 levels are reduced via a proteasome-mediated mechanism. This loss was sustained, as virus replicated in infected cells held at the G(2)/M border. Addition of the cyclin-dependent kinase (CDK) inhibitor roscovitine after S-phase entry reduced MVM replication, suggesting that CDK activity was critical for continued viral replication and virus-induced reduction of p21 may thus be necessary to prevent inhibition of CDK.