EMAIL THIS PAGE TO A FRIEND
Accounts of chemical research
Low-oxidation state indium-catalyzed C-C bond formation.
PMID 22626010
Abstract
The development of innovative metal catalysis for selective bond formation is an important task in organic chemistry. The group 13 metal indium is appealing for catalysis because indium-based reagents are minimally toxic, selective, and tolerant toward various functional groups. Among elements in this group, the most stable oxidation state is typically +3, but in molecules with larger group 13 atoms, the chemistry of the +1 oxidation state is also important. The use of indium(III) compounds in organic synthesis has been well-established as Lewis acid catalysts including asymmetric versions thereof. In contrast, only sporadic examples of the use of indium(I) as a stoichiometric reagent have been reported: to the best of our knowledge, our investigations represent the first synthetic method that uses a catalytic amount of indium(I). Depending on the nature of the ligand or the counteranion to which it is coordinated, indium(I) can act as both a Lewis acid and a Lewis base because it has both vacant p orbitals and a lone pair of electrons. This potential ambiphilicity may offer unique reactivity and unusual selectivity in synthesis and may have significant implications for catalysis, particularly for dual catalytic processes. We envisioned that indium(I) could be employed as a metallic Lewis base catalyst to activate Lewis acidic boron-based pronucleophiles for selective bond formation with suitable electrophiles. Alternatively, indium(I) could serve as an ambiphilic catalyst that activates both reagents at a single center. In this Account, we describe the development of low-oxidation state indium catalysts for carbon-carbon bond formation between boron-based pronucleophiles and various electrophiles. We discovered that indium(I) iodide was an excellent catalyst for α-selective allylations of C(sp(2)) electrophiles such as ketones and hydrazones. Using a combination of this low-oxidation state indium compound and a chiral semicorrin ligand, we developed catalytic highly enantioselective allylation, crotylation, and α-chloroallylation reactions of hydrazones. These transformations proceeded with rare constitutional selectivities and remarkable diastereoselectivities. Furthermore, indium(I) triflate served as the most effective catalyst for allylations and propargylations of C(sp(3)) electrophiles such as O,O-acetals, N,O-aminals, and ethers, and we applied this methodology to carbohydrate chemistry. In addition, a catalyst system composed of indium(I) chloride and a chiral silver BINOL-phosphate facilitated the highly enantioselective allylation and allenylation of N,O-aminals. Overall, these discoveries demonstrate the versatility, efficiency, and sensitivity of low-oxidation state indium catalysts in organic synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57083
Indium, bars, 99.95%
In
264113
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
In
326607
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
In
357278
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
In
326631
Indium, foil, thickness 0.5 mm, 99.999% trace metals basis
In
357286
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
In
264040
Indium, foil, thickness 0.25 mm, 99.998% trace metals basis
In
357294
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
In
264059
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
In
357308
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
In
326615
Indium, pieces, 99.99% trace metals basis
In
203432
Indium, powder, −60 mesh, 99.999% trace metals basis
In
264032
Indium, powder, −100 mesh, 99.99% trace metals basis
In
277959
Indium, powder, 99.99% trace metals basis
In
385379
Indium, rod, diam. 10 mm, 99.9998% trace metals basis
In
264091
Indium, rod, diam. 6.0 mm, ≥99.999% trace metals basis
In
357065
Indium, wire, diam. 2.0 mm, 99.999% trace metals basis
In
278319
Indium, wire, diam. 2.0 mm, 99.99% trace metals basis
In
340863
Indium, wire, diam. 1.5 mm, 99.99% trace metals basis
In
357073
Indium, wire, diam. 1.0 mm, 99.999% trace metals basis
In
264075
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
In
264067
Indium, wire, diam. 0.5 mm, 99.999% trace metals basis
In
326623
Indium, wire, diam. 0.25 mm, 99.999% trace metals basis
In
GF57106403
Indium, foil, 4mm disks, thickness 0.015mm, 99.8%
In
GF38968666
Indium, foil, 4mm disks, thickness 0.01mm, 99.8%
In
GF29602571
Indium, foil, 4mm disks, thickness 0.025mm, 99.8%
In
GF49944088
Indium, foil, 4mm disks, thickness 0.03mm, 99.8%
In
GF38467301
Indium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.999%
In
GF04865564
Indium, foil, 100x100mm, thickness 0.1mm, as rolled, 100%
In
GF40076276
Indium, foil, 100x100mm, thickness 0.20mm, as rolled, 99.999%
In