EMAIL THIS PAGE TO A FRIEND
Cell death & disease
A novel role for RIP1 kinase in mediating TNFα production.
PMID 22695613
Abstract
Receptor-interacting protein 1 (RIP1) is a Ser/Thr kinase with both kinase-dependent and kinase-independent roles in death receptor signaling. The kinase activity of RIP1 is required for necroptosis, a caspase-independent pathway of programmed cell death. In some cell types, the inhibition of caspases leads to autocrine production of TNFα, which then activates necroptosis. Here, we describe a novel role for RIP1 kinase in regulating TNFα production after caspase inhibition. Caspase inhibitors activate RIP1 kinase and another protein, EDD, to mediate JNK signaling, which stimulates Sp1-dependent transcription of TNFα. This pathway is independent of nuclear factor κB and also occurs after Smac mimetic/IAP antagonist treatment or the loss of TNF receptor-associated factor 2 (Traf2). These findings implicate cIAP1/2 and Traf2 as negative regulators of this RIP1 kinase-dependent TNFα production pathway and suggest a novel role for RIP1 kinase in mediating TNFα production under certain conditions.