PloS one
A microRNA-7 binding site polymorphism in HOXB5 leads to differential gene expression in bladder cancer.
PMID 22768238
Abstract
To investigate the biological function of HOXB5 in human bladder cancer and explore whether the HOXB5 3'-UTR SNP (1010A/G), which is located within the microRNA-7 binding site, was correlated with clinical features of bladder cancer. Expression of HOXB5 in 35 human bladder cancer tissues and 8 cell lines were examined using real-time PCR and immunohistochemistry. Next, we explored the biological function of HOXB5 in vitro using cell proliferation, migration and colony formation assays. Using bioinformatics, a SNP (1010A/G) was found located within the microRNA-7 binding site in the 3'-UTR of HOXB5. Real-time PCR was used to test HOXB5 expression affected by different alleles. Finally, multivariate logistic regression analysis was used to determine the relationship between SNP (1010A/G) frequency and clinical features in 391 cases. HOXB5 was frequently over-expressed both in bladder cancer tissues and cell lines. Inhibition of HOXB5 suppressed the oncogenic function of cancer cells. Next, we demonstrated that a SNP (1010A/G), located within the microRNA-7 binding site in the 3'-UTR of HOXB5, could affect HOXB5 expression in bladder cancer mainly by differential binding activity of microRNA-7 and SNP-related mRNA stability. Finally, we also showed the frequency of 1010G genotype was higher in cancer group compared to normal controls and correlated with the risk of high grade and high stage. HOXB5 is overexpressed in bladder cancer. A miRNA-binding SNP (1010A/G) located within 3'-UTR of HOXB5 is associated with gene expression and may be a promising prognostic factor for bladder cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10245 CompoZr® Knockout ZFN Kit, Human HOXB5 (NM_002147)