EMAIL THIS PAGE TO A FRIEND
Organic and medicinal chemistry letters
Discovery of indole tetrafluorophenoxymethylketone-based potent novel small molecule inhibitors of caspase-3.
PMID 22794498
Abstract
Caspase-3 inhibition has been demonstrated to be therapeutically effective in moderating excessive programmed cell death. Interest in caspase-3 as a therapeutic target has led many to pursue the development of inhibitors. To date, only a few series of non-peptide inhibitors have been described, and these have limitations on their drug-like properties. Here, we report the screening of 70 novel small molecules against the caspase-3 enzyme which belongs to four different series (indole fluoromethylketone, indole difluoro and tetrafluorophenoxymethylketone, and oxalamide). Selected molecules were subjected for counter-screening, cell-based, ADME/PK assays in order to understand the potency and drug-like properties. The screening yielded series of hits with IC50 values ranging from 0.11 to 10 μM with reasonable SAR, irreversible mode of inhibition, and reasonable selectivity against other proteases including caspase-1, cathepsin B and D, and thrombin. On the basis of in vitro profile, the selected molecules were evaluated for their drug-like properties. Among the compounds evaluated, compound 3D exhibited good solubility, low permeability, interaction with efflux pump, and low potential for CYP450 drug-drug interaction. After intravenous administration, compound 3D showed low clearance (588 ml/hr/kg), medium volume of distribution, and good oral bioavailability (90%). These results support further advancement of compound 3D in different apoptotic models to develop as a new anti-apoptotic agent in relevant disease conditions.