EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Control of a living radical polymerization of methacrylates by light.
PMID 22807122
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
554065
Ethyl α-bromophenylacetate, 97%
C10H11BrO2