EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Continuous-flow hydrogenation of carbon dioxide to pure formic acid using an integrated scCO2 process with immobilized catalyst and base.
PMID 22807319
Abstract
Dual role for CO(2): Pure formic acid can be obtained continuously by hydrogenation of CO(2) in a single processing unit. An immobilized ruthenium organometallic catalyst and a nonvolatile base in an ionic liquid (IL) are combined with supercritical CO(2) as both reactant and extractive phase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
655422
QuadraPure® TU, macroporous, 400-600 μm particle size