PloS one
Acyl coenzyme A synthetase long-chain 1 (ACSL1) gene polymorphism (rs6552828) and elite endurance athletic status: a replication study.
PMID 22829935
Abstract
The aim of this study was to determine the association between the rs6552828 polymorphism in acyl coenzyme A synthetase (ACSL1) and elite endurance athletic status. We studied 82 Caucasian (Spanish) World/Olympic-class endurance male athletes, and a group of sex and ethnically matched healthy young adults (controls, n=197). The analyses were replicated in a cohort of a different ethnic origin (Chinese of the Han ethnic group), composed of elite endurance athletes (runners) [cases, n=241 (128 male)] and healthy sedentary adults [controls, n=504 (267 male)]. In the Spanish cohort, genotype (P=0.591) and minor allele (A) frequencies were similar in cases and controls (P=0.978). In the Chinese cohort, genotype (P=0.973) and minor allele (G) frequencies were comparable in female endurance athletes and sedentary controls (P=0.881), whereas in males the frequency of the G allele was higher in endurance athletes (0.40) compared with their controls (0.32, P=0.040). The odds ratio (95%CI) for an elite endurance Chinese athlete to carry the G allele compared with ethnically matched controls was 1.381 (1.015-1.880) (P-value=0.04). Our findings suggest that the ACSL1 gene polymorphism rs6552828 is not associated with elite endurance athletic status in Caucasians, yet a marginal association seems to exist for the Chinese (Han) male population.