EMAIL THIS PAGE TO A FRIEND
British dental journal
Buccolam(®) (buccal midazolam): a review of its use for the treatment of prolonged acute convulsive seizures in the dental practice.
PMID 22836423
Abstract
This article is published in response to a query from reader Sarah Clements, based at Golding House Dental Practice Ltd in Kent, regarding the BDJ paper entitled 'Updated guidance on medical emergencies and resuscitation in the dental practice' (BDJ 2012; 212: 41-43). Sarah wished to be provided with further clarification on the ordering, prescribing and administration of midazolam.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M-908
Midazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C18H13ClFN3