EMAIL THIS PAGE TO A FRIEND
The American journal of cardiology
Meta-analysis of safety of the coadministration of statin with fenofibrate in patients with combined hyperlipidemia.
PMID 22840347
Abstract
Addition of fenofibrate to statin therapy might represent a viable treatment option for patients whose high risk for coronary heart disease is not controlled by a statin alone. However, safety of coadministration of statin with fenofibrate has been a great concern. The present study tested the safety of coadministration of statin with fenofibrate. We systematically searched the literature to identify randomized controlled trials examining safety of coadministration of statin with fenofibrate. A meta-analysis was performed to estimate safety of coadministration of statin with fenofibrate using fixed-effects models. There were 1,628 subjects in the identified 6 studies. Discontinuation attributed to any adverse events (4.5% vs 3.1%, p = 0.20), any adverse events (42% vs 41%, p = 0.82), adverse events related to study drug (10.9% vs 11.0%, p = 0.95), and serious adverse events (2.0% vs 1.5%, p = 0.71) were not significantly different in the 2 arms. Incidence of alanine aminotransferase and/or aspartate aminotransferase ≥3 times upper limit of normal in the combination therapy arm was significantly higher than in the statin monotherapy arm (3.1% vs 0.2%, p = 0.0009). In the 6 trials with 1,628 subjects no case of myopathy or rhabdomyolysis was reported. In conclusion, statin-fenofibrate combination therapy was tolerated as well as statin monotherapy. Physicians should consider statin-fenofibrate combination therapy to treat patients with mixed dyslipidemia.
Related Materials