EMAIL THIS PAGE TO A FRIEND
Postgraduate medical journal
Clinical pharmacokinetic studies with lornoxicam.
PMID 2284218
Abstract
Pharmacokinetic data from several studies in young and elderly human subjects are reviewed. Lornoxicam appears to be extensively metabolized and no unchanged drug has been found in the urine. It has a relatively short elimination half-life (about 4 hours), and no significant differences in pharmacokinetic data have been found between young and elderly volunteers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0338
Lornoxicam, ≥98% (HPLC)
C13H10ClN3O4S2