EMAIL THIS PAGE TO A FRIEND
Journal of catalysis
Synthesis, catalytic activity, and leaching studies of a heterogeneous Pd-catalyst including an immobilized bis(oxazoline) ligand.
PMID 22875995
Abstract
The synthesis and characterization of a novel catalytic system including Pd(OAc)(2) attached to a bis(oxazoline) (=BOX) ligand that is covalently bonded to 3-mercaptopropyl-functionalized silica gel is presented. The catalyst was tested for Suzuki-Miyaura reactions of different aryl halides with phenylboronic acid. The heterogeneity of the catalytic system was investigated using different approaches, indicating that there is virtually no Pd leaching into the reaction solution under the applied reaction conditions. Furthermore, our results show that the catalytic system can be reused multiple times without significant loss of stability or structure.
Related Materials