EMAIL THIS PAGE TO A FRIEND
Acta biomaterialia
Cold drawn bioabsorbable ferrous and ferrous composite wires: an evaluation of in vitro vascular cytocompatibility.
PMID 22885027
Abstract
A systematic approach is applied to quantify the impact of bioabsorbable metals on human vascular endothelial cells (EC) and aortic smooth muscle cells (SMC) with the aim of optimizing bioabsorbable endovascular stent development. Composite wires comprising novel combinations of Fe, Mn, Mg, and Zn were produced and fabricated into tubular mesh stents. The stents were incubated with primary EC in order to assess attachment and cell proliferation. Migration of SMCs from the vessel medial wall to the target lesion site following recanalization of an atherosclerotic artery is important in the process of neointimal hyperplasia. Metal ion species were assayed for their impact on cell migration and survival at concentrations ranging from 0.037 to 10 mM. An MTT-based assay was used to assess cytotoxicity after insult with various metal ion concentrations. Fe(2+) and Fe(3+) ion species were found to repress the migration of SMCs across a porous polycarbonate track etch membrane at concentrations of 1 mM. Mn(2+) promoted SMC migration at a concentration of 1 mM, however, this effect was quenched when Fe(2+) was included. Mg(2+) was found to significantly increase SMC migration at concentrations above 1 mM. Cell survival was not reduced after 24 h insult with concentrations of Mg(2+) up to 10 mM. LD50 concentrations of greater than 1 mM were found for Mg(2+), Fe(2+), Fe(3+), and Fe(2+) with 35 wt.% Mn(2+). Significantly greater numbers of EC attached to bioabsorbable metal species compared with 316L stainless steel. Good EC coverage and proliferation were observed for all tested materials up to 120 h.
Related Materials