EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Synthesis of isocyanate-based brush block copolymers and their rapid self-assembly to infrared-reflecting photonic crystals.
PMID 22891697
Abstract
The synthesis of rigid-rod, helical isocyanate-based macromonomers was achieved through the polymerization of hexyl isocyanate and 4-phenylbutyl isocyanate, initiated by an exo-norbornene functionalized half-titanocene complex. Sequential ruthenium-mediated ring-opening metathesis polymerization of these macromonomers readily afforded well-defined brush block copolymers, with precisely tunable molecular weights ranging from high (1512 kDa) to ultrahigh (7119 kDa), while maintaining narrow molecular weight distributions (PDI = 1.08-1.39). The self-assembly of these brush block copolymers to solid thin-films and their photonic properties were investigated. Due to the rigid architecture of these novel polymeric materials, they rapidly self-assemble through simple controlled evaporation to photonic crystal materials that reflect light from the ultra-violet, through the visible, to the near-infrared. The wavelength of reflectance is linearly related to the brush block copolymer molecular weight, allowing for predictable tuning of the band gap through synthetic control of the polymer molecular weight. A combination of scanning electron microscopy and optical modeling was employed to explain the origin of reflectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
414883
Hexyl isocyanate, 97%
C7H13NO