EMAIL THIS PAGE TO A FRIEND
Journal of lipid research
Hyperosmolarity-induced lipid droplet formation depends on ceramide production by neutral sphingomyelinase 2.
PMID 22899568
Abstract
Hyperosmolarity (HO) imposes a remarkable stress on membranes, especially in tissues in direct contact with the external environment. Our efforts were focused on revealing stress-induced lipid changes that precede the inflammatory cytokine response in human corneal epithelial cells exposed to increasing osmolarity. We used a lipidomic analysis that detected significant and systematic changes in the lipid profile, highly correlated with sodium concentrations in the medium. Ceramides and triglycerides (TGs) were the most-responsive lipid classes, with gradual increases of up to 2- and 3-fold, respectively, when compared with control. The source of ceramide proved to be sphingomyelin hydrolysis, and neutral sphingomyelinase 2 (NSM2) activity showed a 2-fold increase 1 h after HO stress, whereas transcription increased 3-fold. Both TG accumulation and IL-8 secretion were shown to be dependent on ceramide production by specific knock-down of NSM2. In HCE cells, diglyceride acyltransferase 1 was responsible for the TG synthesis, but the enzyme activity had no effect on cytokine secretion. Hence, NSM2 plays a key role in the cellular response to hyperosmolar stress, and its activity regulates both cytokine secretion and lipid droplet formation.