EMAIL THIS PAGE TO A FRIEND
Evidence-based complementary and alternative medicine : eCAM
Underlying mechanism of aconitum lizhong acting on experimental hypothermia with indigestion in rats: role of ghrelin.
PMID 22899955
Abstract
This study is aimed to investigate the Aconitum Lizhong pill (ALZ) pharmacological actions on hypothermia with indigestion, especially the ghrelin roles. The littermate-matched rats were randomly divided into four groups. Control did sham operation or standard diet, Model carried out interscapular brown adipose (IBA) removal with standard diet, Fat-diet did IBA removal with fat-diet, and ALZ did IBA removal and fat-diet with 4.536 g/kg/d ALZ. The potency of adaptive thermogenesis, ghrelin levels in plasma or gastric mucosa, thyroid hormones and metabolite in sera, expression of ghrelin mRNA, and protein in gastric mucous membrane were determined. ALZ relieved the hypothermia processes with indigestion, via inhibiting ghrelin expression and increasing ghrelin secretion; the dynamics from the therapy is supported with the energy changes as less body weight loss, less plasma lipid decrease, more plasma T(3) or T(4) increase with TSH decrease, and more compensation of thermogenic AUC decrease. Ghrelin played key roles in the actions of ALZ on the hypothermia with indigestion. The pharmacological mechanisms of ALZ involved the homeostasis of ghrelin expression and secretion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19899
CHAPS 100 mM solution
C32H58N2O7S