EMAIL THIS PAGE TO A FRIEND
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association
What can the dialysis physician learn from kinetic modelling beyond Kt/V(urea)?
PMID 22923544