EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Anti-proliferative effect of methanolic extract of Gracilaria tenuistipitata on oral cancer cells involves apoptosis, DNA damage, and oxidative stress.
PMID 22937998
Abstract
Methanolic extracts of Gracilaria tenuistipitata (MEGT) were obtained from the edible red algae. Previously, we found that water extract of G. tenuistipitata was able to modulate oxidative stress-induced DNA damage and its related cellular responses. In this study, the methanol extraction product MEGT was used to evaluate the cell growth inhibition in oral cancer cells and its possible mechanism was investigated. The cell viability of MEGT treated Ca9-22 oral cancer cell line was significantly decreased in a dose-response manner (p < 0.05). The sub-G1 population and annexin V intensity of MEGT-treated Ca9-22 cancer cells were significantly increased in a dose-response manner (p < 0.0005 and p < 0.001, respectively). The γH2AX intensities of MEGT-treated Ca9-22 cancer cells were significantly increased in a dose-response manner (p < 0.05). The reactive oxygen species (ROS) and glutathione (GSH)-positive intensities of MEGT-treated Ca9-22 oral cancer cells were significantly increased and decreased, respectively, in a dose-response manner (p < 0.05). The DiOC2(3) intensity for mitochondrial membrane potential (MMP) of MEGT-treated Ca9-22 cancer cells was significantly decreased in a dose-response manner (p < 0.05). These results indicated that MEGT had apoptosis-based cytotoxicity against oral cancer cells through the DNA damage, ROS induction, and mitochondrial depolarization. Therefore, MEGT derived from the edible algae may have potential therapeutic effects against oral squamous cell carcinoma (OSCC).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
320684
3,3′-Diethyloxacarbocyanine iodide, 98%
C21H21IN2O2