EMAIL THIS PAGE TO A FRIEND
Medicine, science, and the law
Colchicine poisoning: a case report and review of literature.
PMID 22941521
Abstract
Colchicine, a natural pseudo-alkaloid found in plants such as the Colchicum autumnale and Gloriosa superba has tremendous medicinal properties, but if misused by an unqualified person can result in fatal consequences. We report a case of colchicine poisoning in an adult man as a result of consumption of the herb G. superba by a 50-year-old man and review the literature.