EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Convergent solid-phase synthesis of N-glycopeptides facilitated by pseudoprolines at consensus-sequence Ser/Thr residues.
PMID 22945377
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
802999
2-Acetamido-2-deoxy-β-D-glucopyranosylamine, 97% (HPLC)
C8H16N2O5