PloS one
miR-421 and miR-30c inhibit SERPINE 1 gene expression in human endothelial cells.
PMID 22952991
Abstract
In this work, we assessed whether SERPINE1 expression could be under the influence of microRNAs (miRNAs) predicted to bind the SERPINE1 3'UTR region. We specifically focused on the 3'UTR region harboring a common polymorphism, rs1050955, that have been found associated to SERPINE1 monocyte expression, and investigated whether the presence of different alleles at rs1050955 could modify the miRNAs binding efficiency and affect PAI-1 protein levels. We demonstrated that, in human umbilical vein endothelial cells, both miR-421 and miR-30c directly interacted with PAI-1 mRNA to inhibit the expression of the associated protein. However, these inhibitory mechanisms were independent on the allele present at the rs1050955 locus. We further showed that miR-421 levels correlated with PAI-1 activity in the plasma sample of 40 patients with venous thrombosis. Our results strongly suggest that the regulation of PAI-1 molecule could be under the influence of several miRNAs whose measurement in the plasma of patients could be envisaged as a biomarker for inflammatory and thrombotic disorders.