EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Visible light photocatalysis of [2+2] styrene cycloadditions by energy transfer.
PMID 22965321
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45993
Styrene, analytical standard
C8H8