EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
The need for combined inorganic, biochemical, and nutritional studies of chromium(III).
PMID 22976981
Abstract
The history of biochemical and nutritional studies of the element is unfortunately full of twists and turns, most leading to dead ends. Chromium (Cr), as the trivalent ion, has been proposed to be an essential element, a body mass and muscle development agent, and, in the form of the most popular Cr-containing nutritional supplement, to be toxic when given orally to mammals. None of these proposals, despite significant attention in the popular media, has proven to be correct. Trivalent chromium has also been proposed as a therapeutic agent to increase insulin sensitivity and affect lipid metabolism, although a molecular mechanism for such actions has not been elucidated. Greater cooperative research interactions between nutritionists, biochemists, and chemists might have avoided the earlier issues in nutritional and biochemical Cr research and is necessary to establish the potential role of Cr as a therapeutic agent at a molecular level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF97385338
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF23770885
Chromium, foil, not light tested, 100x100mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF24605816
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF22618739
Chromium, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr
GF00270593
Chromium, foil, not light tested, 25x25mm, thickness 0.0015mm, permanent polyester support, 99.99+%
Cr
GF88955410
Chromium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF94865170
Chromium, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF73692377
Chromium, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF61658725
Chromium, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF81124286
Chromium, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr
GF05241316
Chromium, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.99+%
Cr
GF98123250
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Cr
GF34438923
Chromium, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%
Cr
GF49392456
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Cr
GF88985446
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Cr
GF30096053
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF12477157
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF84397168
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Cr
GF50354692
Chromium, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF74347050
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF07622759
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Cr
GF35390651
Chromium, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF46122645
Chromium, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF31792956
Chromium, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr
GF91240994
Chromium, foil, not light tested, 25x25mm, thickness 0.075mm, permanent polyester support, 99.99+%
Cr
GF98190861
Chromium, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF76896400
Chromium, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF64373766
Chromium, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF34299748
Chromium, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF31161754
Chromium, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr