EMAIL THIS PAGE TO A FRIEND
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova
[Possibilities in using sulpiride in the treatment of psychotic disorders].
PMID 22983256
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S2190000
Sulpiride, European Pharmacopoeia (EP) Reference Standard
C15H23N3O4S