EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Oxaliplatin-induced thrombotic thrombocytopenic purpura: case report and literature review.
PMID 22987080