EMAIL THIS PAGE TO A FRIEND
International journal of technology assessment in health care
Benefits of pharmaceutical innovation: the case of simvastatin in Canada.
PMID 22989373
Abstract
The benefits of pharmaceutical innovations are widely diffused; they accrue to the healthcare providers, patients, employers, and manufacturers. We estimate the societal monetary benefits of simvastatin in Canada and its distribution among different beneficiaries overtime. Monetary benefits to developing and generic manufacturers were estimated by calculating public and private revenues minus the development costs of simvastatin and the contribution toward further research and development. We used a dynamic Markov model to estimate monetary benefits to healthcare and employment sectors in terms of cost avoidance associated with prevented cardiovascular events, including stroke and myocardial infarction, and lost productivity due to disability and premature death in working population. Cumulative monetary benefits of simvastatin from 1990 to 2009 were $4.8 billion (2010 CA$), of which developing and generic manufacturers, and healthcare and employment sectors accounted for 32 percent, 27 percent, 32 percent, and 9 percent, respectively. The yearly trend showed that after the patent expired in 2002 the generic manufacturers became dominant in the market. Benefits for the healthcare sector started to decrease from 2003 corresponding to the decreasing population taking simvastatin during the same time period. Sensitivity analysis showed the higher the compliance or the efficacy, the larger the benefits to healthcare and employment sectors, while monetary benefits for manufacturers were unchanged. Societal monetary benefits of simvastatin are significant and the distributions of the benefits have changed overtime. Patent, compliance, and efficacy play a vital role in the estimation of the benefits. Analysis of all beneficiaries separately overtime is important when assessing the value of pharmaceutical innovation.