EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
Parallel solid-phase synthesis of diaryltriazoles.
PMID 23019430
Abstract
A series of substituted diaryltriazoles was prepared by a solid-phase-synthesis protocol using a modified Wang resin. The copper(I)- or ruthenium(II)-catalyzed 1,3-cycloaddition on the polymer bead allowed a rapid synthesis of the target compounds in a parallel fashion with in many cases good to excellent yields. Substituted diaryltriazoles resemble a molecular structure similar to established terphenyl-alpha-helix peptide mimics and have therefore the potential to act as selective inhibitors for protein-protein interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
557331
3-Ethynyl-α,α,α-trifluorotoluene, 97%
C9H5F3