EMAIL THIS PAGE TO A FRIEND
Internal and emergency medicine
Helicobacter pylori secreted peptidyl prolyl cis, trans-isomerase drives Th17 inflammation in gastric adenocarcinoma.
PMID 23054412
Abstract
Helicobacter pylori infection is characterized by an inflammatory infiltrate, consisting mainly of neutrophils and T cells. This study was undertaken to evaluate the type of gastric T cell response elicited by the secreted peptidyl prolyl cis, trans-isomerase of H. pylori (HP0175) in patients with distal gastric adenocarcinoma. The cytokine profile and the effector functions of gastric tumor-infiltrating lymphocytes (TILs) specific for HP0175 was investigated in 20 patients with distal gastric adenocarcinoma and H. pylori infection. The helper function of HP0175-specific TILs for monocyte MMP-2, MMP-9, and VEGF production was also investigated. TILs cells from H. pylori infected patients with distal gastric adenocarcinoma produced Interleukin (IL)-17 and IL-21 in response to HP0175. HP0175-specific TILs showed poor cytolytic activity while expressing helper activity for monocyte MMP-2, MMP-9 and VEGF production. These findings indicate that HP0175 is able to drive gastric Th17 response. Thus, HP0175, by promoting pro-inflammatory low cytotoxic TIL response, matrix degradation and pro-angiogenic pathways, may provide a link between H. pylori and gastric cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0037
Human Procalcitonin ELISA Kit