EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Suppression and recovery of BRCA1-mediated transcription by HP1γ via modulation of promoter occupancy.
PMID 23074186
Abstract
Heterochromatin protein 1γ (HP1γ) is a chromatin protein involved in gene silencing. Herein, we show that HP1γ interacts with breast cancer type 1 susceptibility protein (BRCA1) and regulates BRCA1-mediated transcription via modulation of promoter occupancy and histone modification. We used several HP1γ mutants and small interfering RNAs for histone methyltransferases to show that BRCA1-HP1γ interaction, but not methylated histone binding, is important in HP1γ repression of BRCA1-mediated transcription. Time-lapse studies on promoter association and histone methylation after DNA damage revealed that HP1γ accumulates at the promoter before DNA damage, but BRCA1 is recruited at the promoter after the damage while promoter-resident HP1γ is disassembled. Importantly, HP1γ assembly recovers after release from the damage in a BRCA1-HP1γ interaction-dependent manner and targets SUV39H1. HP1γ/SUV39H1 restoration at the promoter results in BRCA1 disassembly and histone methylation, after which transcription repression resumes. We propose that through interaction with BRCA1, HP1γ is guided to the BRCA1 target promoter during recovery and functions in the activation-repression switch and recovery from BRCA1-mediated transcription in response to DNA damage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-942
Anti-acetyl-Histone H3 (Lys9) Antibody, 200 µg, Anti-acetyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 acetylated on lysine 9. This purified Ab, also known as Anti-H3K9Ac, is specificity verified by dot blot (DB), published in peer reviewed journals, and validated in WB, ChIP, ChIP-seq, FC.
MAB3448
Anti-Heterochromatin Protein-1 β Antibody, clone 1MOD-1A9, 100 µL, Use Anti-Heterochromatin Protein-1 β Antibody, clone 1MOD-1A9 (Mouse Monoclonal Antibody) validated in ELISA, ICC, IHC, IP, WB to detect Heterochromatin Protein-1 β also known as HP1b.
MAB3584
Anti-Heterochromatin Protein-1 α Antibody, clone 2HP-1H5, 100 µL, Use Anti-Heterochromatin Protein-1 α Antibody, clone 2HP-1H5 (mouse monoclonal antibody) validated in ELISA, ICC, IHC to detect Heterochromatin Protein-1 α also known as HP1a, Antigen p25, Chromobox Protein Homolog 5.
MAB3450
Anti-Heterochromatin Protein-1 gamma Antibody, clone 2MOD-1G6, 100 µL, Anti-Heterochromatin Protein-1 gamma Antibody, clone 2MOD-1G6 is a Mouse Monoclonal Antibody for detection of Heterochromatin Protein-1 gamma also known as HP-1gamma & has been validated in ELISA, ICC, IF, IHC & WB.
05-690
Anti-HP1γ Antibody, clone 42s2, 200 µg, Use Anti-HP1γ Antibody, clone 42s2 (Mouse Monoclonal Antibody) validated in ChIP, IHC, WB to detect HP1γ also known as Heterochromatin protein 1 homolog gamma.
06-719
Anti-LexA Antibody, DNA-binding region, 200 µg, Detect LexA with Anti-LexA Antibody, DNA-binding region (Rabbit Polyclonal Antibody), that has been shown to work in IP & WB.
07-442
Anti-trimethyl-Histone H3 (Lys9) Antibody, 100 µg, Use Anti-trimethyl-Histone H3 (Lys9) Antibody (rabbit polyclonal antibody) validated in DB, ICC, Mplex, PIA, WB, ChIP-seq to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).