EMAIL THIS PAGE TO A FRIEND
Toxicology and industrial health
Mitigation of hepatotoxic effects of arsenic trioxide through omega-3 fatty acid in rats.
PMID 23081861
Abstract
Arsenic trioxide (As(2)O(3)) is an effective drug in the treatment of leukaemia and many solid tumours. In clinical trials, arsenic therapy is closely associated with hepatic toxicity. The present study was designed to investigate the efficacy of omega-3 fatty acid against As(2)O(3)-induced hepatotoxicity. A 4 mg/kg body weight (bw) of As(2)O(3) was orally administered to Wistar male rats for 45 days. Hepatotoxicity was evaluated by biochemical tests, antioxidant assays and histopathological examinations. Arsenic accumulation was found in the liver tissue of rats treated with As(2)O(3). Hepatoprotective efficacy of omega-3 fatty acid was analysed by the combination therapy with As(2)O(3). In vivo studies revealed a significant rise in lipid peroxidation with concomitant decline in reduced glutathione, glutathione-dependant antioxidant enzymes and antiperoxidative enzymes in the liver tissue of rats treated with arsenic. The supplementation of omega-3 fatty acid at a dose of 50 mg/kg bw with As(2)O(3) offers ameliorative effect against hepatocellular toxicity. Omega-3 fatty acid maintained hepatic marker enzymes, antioxidant enzymes and decreased lipid peroxidation. The combination treatment clearly reduced the hepatic structural abnormalities such as haemorrhage, necrosis and cholangiofibrosis in the rats treated with arsenic. This study concludes that the omega-3 fatty acid might be useful for the protection against As(2)O(3)-induced hepatotoxicity.